α,α'-Dichlor-p-xylol
Lieferant: Molekula
Synonyme:
1,4-Bis(chlormethyl)benzol, p-Xyloldichlorid, p-Xylylendichlorid
38947698-100G
MOLE38947698-100GEA
128
EUR
MOLE38947698-100G
α,α'-Dichlor-p-xylol
α,α'-Dichlor-p-xylol
Formel:
C₈H₈Cl₂ Molecular Weight: 175,06 g/mol Siedepunkt: 254 °C (1013 hPa) Schmelzpunkt: 100 °C Dichte: 1,432 g/cm³ (20 °C) Lagertemperatur: Kühlschrank |
MDL:
MFCD00000920 CAS-Nummer: 623-25-6 EINECS: 210-782-7 UN: 2928 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...