TBTU (O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluroniumtetrafluoroborat)
Lieferant: Molekula
Synonyme:
TBTU
39093565-5G
MOLE39093565-5GEA
0
EUR
MOLE39093565-5G
TBTU (O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluroniumtetrafluoroborat)
TBTU (O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluroniumtetrafluoroborat)
Formel:
C₁₁H₁₆BF₄N₅O Molecular Weight: 321,1 g/mol |
MDL:
MFCD00077413 CAS-Nummer: 125700-67-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...