(±)-Taxifolin

Lieferant: Molekula

Synonyme: Taxifolin, Dihydroquercetin, 3,3′,4′,5,7-Pentahydroxyflavanone

39141540-100MG
MOLE39141540-100MGEA 0 EUR
MOLE39141540-100MG
(±)-Taxifolin
(±)-Taxifolin
Formel: C₁₅H₁₂O₇
Molecular Weight: 304,25 g/mol (anhy)
MDL: MFCD11044454
CAS-Nummer: 24198-97-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR