α-Brom-3-iodtoluol
Lieferant: Molekula
|
Gefahr
|
Synonyme:
α-Brom-m-iodtoluol, 3-Iodbenzylbromid
39198942-5G
MOLE39198942-5GEA
147
EUR
MOLE39198942-5G
α-Brom-3-iodtoluol
α-Brom-3-iodtoluol
Formel:
C₇H₆BrI Molecular Weight: 296,93 g/mol Schmelzpunkt: 50…52 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00019016 CAS-Nummer: 49617-83-6 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...