4-Fluorindol

Lieferant: Molekula
Achtung

Synonyme: 4-Fluoro-1H-indole

39257867-1G
MOLE39257867-1GEA 71.6 EUR
MOLE39257867-1G
4-Fluorindol
4-Fluorindol
Formel: C₈H₆FN
Molecular Weight: 135.14 g/mol
MDL: MFCD00055992
CAS-Nummer: 387-43-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR