4-Chlor-4'-hydroxybenzophenon

Lieferant: Molekula

Synonyme: (4-Chlorophenyl)(4-hydroxyphenyl)methanone

39582283-100G
MOLE39582283-100GEA 0 EUR
MOLE39582283-100G
4-Chlor-4'-hydroxybenzophenon
4-Chlor-4'-hydroxybenzophenon
Formel: C₁₃H₉ClO₂
MDL: MFCD00002357
CAS-Nummer: 42019-78-3
EINECS: 255-627-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR