2-Chloroethanesulphonic acid sodium salt hydrate
Lieferant: Molekula
Synonyme:
Sodium-2-chloroethanesulphonate hydrate
39693672-25G
MOLE39693672-25GEA
0
EUR
MOLE39693672-25G
2-Chloroethanesulphonic acid sodium salt hydrate
2-Chloroethanesulphonic acid sodium salt hydrate
Formel:
C₂H₄ClNaO₃S.H₂O Molecular Weight: 184,57 g/mol |
MDL:
MFCD00007531 CAS-Nummer: 1089719-72-1 EINECS: 241-935-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...