2-Benzofurancarbonsäure

Lieferant: Molekula

Synonyme: Benzo[b]furan-2-carboxylic acid

39760752-5G
MOLE39760752-5GEA 0 EUR
MOLE39760752-5G
2-Benzofurancarbonsäure
Benzofuran-2-carboxylic acid
Formel: C₉H₆O₃
Molecular Weight: 162.15 g/mol
MDL: MFCD00005848
CAS-Nummer: 496-41-3
EINECS: 207-818-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR