DL-α-Tocopherol
Lieferant: Molekula
Synonyme:
(±)-α-Tocopherol, all-rac-α-Tocopherol, 3,4-Dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-2H-benzopyran-6-ol, Vitamin E Alkohol, Vitamin E
39762555-100G
MOLE39762555-100GEA
0
EUR
MOLE39762555-100G
DL-α-Tocopherol
DL-α-Tocopherol
Formel:
C₂₉H₅₀O₂ Molecular Weight: 430,71 g/mol |
MDL:
MFCD00072051 CAS-Nummer: 10191-41-0 EINECS: 233-466-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...