2-(2-Fluorphenyl)ethanol

Lieferant: Molekula

Synonyme: 2-Fluorophenethyl alcohol

39983774-5G
MOLE39983774-5GEA 0 EUR
MOLE39983774-5G
2-(2-Fluorphenyl)ethanol
2-(2-Fluorphenyl)ethanol
Formel: C₈H₉FO
Molecular Weight: 140.16 g/mol
Siedepunkt: 104…106 °C (14 mmHg)
Dichte: 1.04 g/cm³ (20 °C)
MDL: MFCD00002887
CAS-Nummer: 50919-06-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR