Benzo[b]furan-2-boronsäure
Lieferant: Molekula
Synonyme:
Benzofuran-2-boronic acid, 2-Benzofurylboronic acid
40173470-1G
MOLE40173470-1GEA
0
EUR
MOLE40173470-1G
Benzo[b]furan-2-boronsäure
Benzo[b]furan-2-boronsäure
Formel:
C₈H₇BO₃ Molecular Weight: 161,95 g/mol |
MDL:
MFCD00236019 CAS-Nummer: 98437-24-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...