Adeninsulfat

Lieferant: Molekula

40343098-100G
MOLE40343098-100GEA 0 EUR
MOLE40343098-100G
Adeninsulfat
Adeninsulfat
Formel: C₅H₅N₅·½H₂SO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00213655
CAS-Nummer: 321-30-2
EINECS: 206-286-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR