D(+)-Galactosamin Hydrochlorid

Lieferant: Molekula

Synonyme: (2R,3R,4R,5R)-2-Amino-3,4,5,6-tetrahydroxyhexanal hydrochloride

40622754-5G
MOLE40622754-5GEA 86 EUR
MOLE40622754-5G
D(+)-Galactosamin Hydrochlorid
D(+)-Galactosamin Hydrochlorid
Formel: C₆H₁₄ClNO₅
Molecular Weight: 215,64 g/mol
MDL: MFCD00135830
CAS-Nummer: 1772-03-8
EINECS: 217-198-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR