2,4-Dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamid
Lieferant: Molekula
Synonyme:
Panthenol, DL-Pantothenol, 2,4-Dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamide
41027803-100G
MOLE41027803-100GEA
101
EUR
MOLE41027803-100G
2,4-Dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamid
DL-Panthenol
Formel:
C₉H₁₉NO₄ Molecular Weight: 205.25 g/mol |
MDL:
MFCD00002944 CAS-Nummer: 16485-10-2 EINECS: 240-540-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...