Thiophen-2-ethylamin
Lieferant: Molekula
Synonyme:
2-(Thiophen-2-yl)ethanamine, 2-(2-Aminoethyl)thiophene
41164148-1G
MOLE41164148-1GEA
0
EUR
MOLE41164148-1G
Thiophen-2-ethylamin
Thiophen-2-ethylamin
Formel:
C₆H₉NS Molecular Weight: 127,21 g/mol |
MDL:
MFCD00051495 CAS-Nummer: 30433-91-1 EINECS: 250-196-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...