4-(Methylamino)pyridin

Lieferant: Molekula

41435257-5G
MOLE41435257-5GEA 0 EUR
MOLE41435257-5G
4-(Methylamino)pyridin
4-(Methylamino)pyridine
Formel: C₆H₈N₂
Molecular Weight: 108.14 g/mol
MDL: MFCD00152625
CAS-Nummer: 1121-58-0
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR