Ketoprofen

Lieferant: Molekula

Synonyme: 2-(3-Benzoylphenyl)propionic acid, 2-(3-Benzoylphenyl)propanoic acid

41476439-5G
MOLE41476439-5GEA 0 EUR
MOLE41476439-5G
Ketoprofen
Ketoprofen
Formel: C₁₆H₁₄O₃
MDL: MFCD00055790
CAS-Nummer: 22071-15-4
EINECS: 244-759-8
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR