Benzo-18-krone-6 (Benzo-1,4,7,10,13,16-hexoxacyclooctadecan)
Lieferant: Molekula
Synonyme:
Benzo-18-crown 6-Ether
41515582-1G
MOLE41515582-1GEA
0
EUR
MOLE41515582-1G
Benzo-18-krone-6 (Benzo-1,4,7,10,13,16-hexoxacyclooctadecan)
Benzo-18-krone-6 (Benzo-1,4,7,10,13,16-hexoxacyclooctadecan)
Formel:
C₁₆H₂₄O₆ Molecular Weight: 312.36 g/mol |
MDL:
MFCD00062741 CAS-Nummer: 14098-24-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...