4-(Trifluormethyl)phenylhydrazin

Lieferant: Molekula

41639505-5G
MOLE41639505-5GEA 0 EUR
MOLE41639505-5G
4-(Trifluormethyl)phenylhydrazin
4-(Trifluormethyl)phenylhydrazin
Formel: C₇H₇F₃N₂
MDL: MFCD00042508
CAS-Nummer: 368-90-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR