Benzo-12-krone-4 (Benzo-1,4,7,10-tetraoxacyclododecan)

Lieferant: Molekula

Synonyme: Benzo-12-crown 4-Ether, 2,3-Benzo-1,4,7,10-tetraoxadodec-2-ene, Benzo-12-crown-4

41748345-1G
MOLE41748345-1GEA 0 EUR
MOLE41748345-1G
Benzo-12-krone-4 (Benzo-1,4,7,10-tetraoxacyclododecan)
Benzo-12-krone-4 (Benzo-1,4,7,10-tetraoxacyclododecan)
Formel: C₁₂H₁₆O₄
Molecular Weight: 224.26 g/mol
MDL: MFCD00059934
CAS-Nummer: 14174-08-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR