4-Chlor-3-nitrobenzylalkohol

Lieferant: Molekula

41754298-25G
MOLE41754298-25GEA 0 EUR
MOLE41754298-25G
4-Chlor-3-nitrobenzylalkohol
4-Chlor-3-nitrobenzylalkohol
Formel: C₇H₆ClNO₃
Schmelzpunkt: 66 °C
MDL: MFCD00007086
CAS-Nummer: 55912-20-4
EINECS: 259-901-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR