γ-Cyclodextrin
Lieferant: Molekula
Synonyme:
gamma-Cyclodextrin, Cyclooctaamylose, γ-Cyclodextrin, Schardinger gamma-Dextrin
41851510-1G
MOLE41851510-1GEA
219
EUR
MOLE41851510-1G
γ-Cyclodextrin
γ-Cyclodextrin
Formel:
C₄₈H₈₀O₄₀ Molecular Weight: 1297,1248 g/mol |
MDL:
MFCD00009595 CAS-Nummer: 17465-86-0 EINECS: 241-482-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...