N-Octadecyl-N,N-dimethyl-3-ammonio-1-propansulfonat (Sulfobetain-18, SB-18)

Lieferant: Molekula

Synonyme: Stearyldimethyl(3-sulphopropyl)ammonium Hydroxide Inner salt, 3-[Dimethyl(octadecyl)ammonio]propane-1-sulphonate, Stearyl sulphobetaine, Octadecyldimethyl(3-sulphopropyl)ammonium Hydroxide Inner salt, 3-[Dimethyl(stearyl)ammonio]propane-1-sulphonate

42559860-25G
MOLE42559860-25GEA 0 EUR
MOLE42559860-25G
N-Octadecyl-N,N-dimethyl-3-ammonio-1-propansulfonat (Sulfobetain-18, SB-18)
N-Octadecyl-N,N-dimethyl-3-ammonio-1-propansulfonat (Sulfobetain-18, SB-18)
Formel: C₂₃H₄₉NO₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD00043218
CAS-Nummer: 13177-41-8
EINECS: 236-124-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR