Orange G
Lieferant: Molekula
Synonyme:
Acid Orange 10, Dinatrium-7-hydroxy-8-phenylazonaphthalin-1,3-disulfonat, Orange G Natriumsalz, CI 16230
42617597-25G
MOLE42617597-25GEA
101
EUR
MOLE42617597-25G
Orange G
Orange G
Formel:
C₁₆H₁₀N₂Na₂O₇S₂ Molecular Weight: 452,38 g/mol Schmelzpunkt: 141 °C Dichte: 0,45…0.47 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012457 CAS-Nummer: 1936-15-8 EINECS: 217-705-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...