4-Nitro-o-phenylendiamin

Lieferant: Molekula
Achtung

42675126
MOLE42675126EA 0 EUR
MOLE42675126
4-Nitro-o-phenylendiamin
4-Nitro-o-phenylendiamin
Formel: C₆H₇N₃O₂
Schmelzpunkt: 197…202 °C
MDL: MFCD00007724
CAS-Nummer: 99-56-9
EINECS: 202-766-3
Merck Index: 12,06720

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR