(S)-(-)-1,2,3,4-Tetrahydroisochinolin-3-carbonsäure
Lieferant: Molekula
Synonyme:
(3S)-1,2,3,4-Tetrahydro-3-isoquinolinecarbocylic acid, (S)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid
42731182-5G
MOLE42731182-5GEA
0
EUR
MOLE42731182-5G
(S)-(-)-1,2,3,4-Tetrahydroisochinolin-3-carbonsäure
(S)-(-)-1,2,3,4-Tetrahydroisochinolin-3-carbonsäure
Formel:
C₁₀H₁₁NO₂ Molecular Weight: 177,20 g/mol |
MDL:
MFCD00144533 CAS-Nummer: 74163-81-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...