DL-Cystein
Lieferant: Molekula
Synonyme:
H-DL-Cys-OH, (±)-2-Amino-3-mercaptopropansäure, (±)-Cystein
42875416
MOLE42875416EA
139
EUR
MOLE42875416
DL-Cystein
DL-Cystein
Formel:
C₃H₇NO₂S Molecular Weight: 121,16 g/mol Siedepunkt: 254 °C (1013 hPa) Schmelzpunkt: 115 °C Dichte: 1,441 g/cm³ (25 °C) |
MDL:
MFCD00004881 CAS-Nummer: 3374-22-9 EINECS: 222-160-2 Merck Index: 12,02850 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...