(S)-(+)-1,2-Propandiol
Lieferant: Molekula
Synonyme:
(S)-(+)-1,2-Dihydroxypropan, (S)-(+)-1,2-Propylenglykol, (S)-(+)-1,2-Propylenglycol
43121408-1G
MOLE43121408-1GEA
89.7
EUR
MOLE43121408-1G
(S)-(+)-1,2-Propandiol
(S)-(+)-1,2-Propandiol
Formel:
C₃H₈O₂ Molecular Weight: 76,1 g/mol Dichte: 1,03 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004539 CAS-Nummer: 4254-15-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...