6-O-Palmitoyl-L-ascorbinsäure

Lieferant: Molekula

Synonyme: 6-palmitoyl-L-ascorbic acid, L(+)-Ascorbylpalmitat

43238027-100G
MOLE43238027-100GEA 85.7 EUR
MOLE43238027-100G
6-O-Palmitoyl-L-ascorbinsäure
6-O-Palmitoyl-L-ascorbinsäure
Formel: C₂₂H₃₈O₇
Molecular Weight: 414,54 g/mol
Siedepunkt: ∼250 °C
Schmelzpunkt: 107…117 °C
MDL: MFCD00005377
CAS-Nummer: 137-66-6
EINECS: 205-305-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR