2-Fluor-4-nitrophenol

Lieferant: Molekula

43304392-5G
MOLE43304392-5GEA 0 EUR
MOLE43304392-5G
2-Fluor-4-nitrophenol
2-Fluor-4-nitrophenol
Formel: C₆H₄FNO₃
Molecular Weight: 157,1 g/mol
Schmelzpunkt: 118…118,5 °C
MDL: MFCD00051970
CAS-Nummer: 403-19-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR