2-Mercapto-5-methylbenzimidazol
Lieferant: Molekula
Synonyme:
2-Mercapto-5-methyl-1H-benzimidazol, 1,3-Dihydro-5-methyl-2H-benzimidazol-2-thion
43308264-100G
MOLE43308264-100GEA
0
EUR
MOLE43308264-100G
2-Mercapto-5-methylbenzimidazol
2-Mercapto-5-methylbenzimidazole
Formel:
C₈H₈N₂S Molecular Weight: 164,23 g/mol Siedepunkt: 349 °C (1013 hPa) Schmelzpunkt: 220 °C |
MDL:
MFCD00010617 CAS-Nummer: 27231-36-3 EINECS: 248-350-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...