Sudan III

Lieferant: Molekula

Synonyme: Sudan Rot, Sudan Red BK, Sudan® III (Rot), 1-(4-(Phenylazo)phenylazo)-2-naphthol, Tony Red, Scarlet B, 1-([4-[(Phenyl)diazenyl]phenyl]diazenyl)naphthalen-2-ol, Fat Ponceau G, Sudan G, Cerasin Red, Sudan Red B, Organol® Rot BS, Sudan Red III, Sudan V

43449981-25G
MOLE43449981-25GEA 0 EUR
MOLE43449981-25G
Sudan III
Sudan III
Formel: C₂₂H₁₆N₄O
Molecular Weight: 352,4 g/mol
Schmelzpunkt: 205,3…206.3 °C
MDL: MFCD00003905
CAS-Nummer: 85-86-9
EINECS: 201-638-4
Merck Index: 12,09054

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR