2-Cyan-3-methylanilin

Lieferant: Molekula

Synonyme: 2-Cyano-3-methylaniline

43603587-5G
MOLE43603587-5GEA 0 EUR
MOLE43603587-5G
2-Cyan-3-methylanilin
2-Cyan-3-methylanilin
Formel: C₈H₈N₂
Molecular Weight: 132,16 g/mol
MDL: MFCD00058686
CAS-Nummer: 56043-01-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR