FITC (Fluorescein-5-isothiocyanat, Fluoresceinisothiocyanat Isomer I)

Lieferant: Molekula
Achtung

43825432-1G
MOLE43825432-1GEA 0 EUR
MOLE43825432-1G
FITC (Fluorescein-5-isothiocyanat, Fluoresceinisothiocyanat Isomer I)
FITC (Fluorescein-5-isothiocyanat, Fluoresceinisothiocyanat Isomer I)
Formel: C₂₁H₁₁NO₅S
Lagertemperatur: Kühlschrank
MDL: MFCD00005063
CAS-Nummer: 3326-32-7
EINECS: 222-042-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR