Ethyl-5-amino-1-phenyl-1H-pyrazol-4-carboxylat

Lieferant: Molekula

Synonyme: 5-Amino-1-phenylpyrazole-4-carboxylic acid Ethyl Ester, 5-Amino-1-phenyl-1H-pyrazole-4-carboxylic acid ethyl ester, 5-Amino-1-phenylpyrazole-4-carboxylic acid ethyl ester, 5-Amino-4-carbethoxy-1-phenylpyrazole, Ethyl 5-Amino-1-phenylpyrazole-4-carboxylate, Ethyl 5-amino-1-phenyl-1H-pyrazole-4-carboxylate

43889017-5G
MOLE43889017-5GEA 0 EUR
MOLE43889017-5G
Ethyl-5-amino-1-phenyl-1H-pyrazol-4-carboxylat
Ethyl-5-amino-1-phenyl-1H-pyrazol-4-carboxylat
Formel: C₇H₁₁N₃O₂
Molecular Weight: 231.26 g/mol
MDL: MFCD00020731
CAS-Nummer: 16078-71-0
EINECS: 240-224-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR