2-Methylbenzofuran

Lieferant: Molekula

43971397-1G
MOLE43971397-1GEA 362 EUR
MOLE43971397-1G
2-Methylbenzofuran
2-Methylbenzofuran
Formel: C₉H₈O
Molecular Weight: 132,16 g/mol
MDL: MFCD00005850
CAS-Nummer: 4265-25-2
EINECS: 224-249-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR