D(+)-Raffinose Pentahydrat
Lieferant: Molekula
Synonyme:
Melitose Pentahydrat, Melitriose Pentahydrat, Gossypose Pentahydrat
44163104-25G
MOLE44163104-25GEA
0
EUR
MOLE44163104-25G
D(+)-Raffinose Pentahydrat
D(+)-Raffinose Pentahydrat
Formel:
C₁₈H₃₂O₁₆·5H₂O Molecular Weight: 594,52 g/mol Schmelzpunkt: 80 °C Dichte: 1,516 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00071590 CAS-Nummer: 17629-30-0 EINECS: 208-146-9 Merck Index: 12,08279 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...