m-Toluonitril
Lieferant: Molekula
Synonyme:
3-Methylbenzoesäurenitril, 3-Methylbenzonitril, m-Methylbenzonitril
44329754-100G
MOLE44329754-100GEA
0
EUR
MOLE44329754-100G
m-Toluonitril
m-Toluonitril
Formel:
C₈H₇N Molecular Weight: 117,15 g/mol Siedepunkt: 213 °C (1013 hPa) Schmelzpunkt: –0,5 °C Dichte: 0,976 g/cm³ (20 °C) |
MDL:
MFCD00001808 CAS-Nummer: 620-22-4 EINECS: 210-631-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...