(-)-Di-p-toluoyl-L-weinsäure
Lieferant: Molekula
Synonyme:
Di-p-toluoyl-L-tartaric acid, (-)-O,O'-Di-p-toluoyl-L-tartaric acid, (2R,3R)-(-)-Di-O-4-Toluoyl-L-Tartaric Acid
44432850-25G
MOLE44432850-25GEA
0
EUR
MOLE44432850-25G
(-)-Di-p-toluoyl-L-weinsäure
(-)-Di-p-toluoyl-L-weinsäure
Formel:
C₂₀H₁₈O₈ Molecular Weight: 386.36 g/mol |
MDL:
MFCD00064440 CAS-Nummer: 32634-66-5 EINECS: 251-131-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...