Tetrabutylammoniumchlorid 50% in wässriger Lösung
Lieferant: Molekula
44513467-100ML
MOLE44513467-100MLEA
0
EUR
MOLE44513467-100ML
Tetrabutylammoniumchlorid 50% in wässriger Lösung
Tetrabutylammoniumchlorid
Formel:
C₁₆H₃₆ClN Molecular Weight: 277,92 g/mol |
MDL:
MFCD00011635 CAS-Nummer: 1112-67-0 EINECS: 214-195-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...