1-Aza-18-krone-6 (1,4,7,10,13-Pentaoxa-16-azacyclooctadecan)

Lieferant: Molekula

Synonyme: 1,4,7,10,13-Pentaoxa-16-azacyclooctadecane, 1-Aza-18-crown 6-Ether, 1-Aza-18-crown-6

44608555-1G
MOLE44608555-1GEA 315 EUR
MOLE44608555-1G
1-Aza-18-krone-6 (1,4,7,10,13-Pentaoxa-16-azacyclooctadecan)
1-Aza-18-krone-6 (1,4,7,10,13-Pentaoxa-16-azacyclooctadecan)
Formel: C₁₂H₂₅NO₅
Molecular Weight: 263.33 g/mol
MDL: MFCD00075466
CAS-Nummer: 33941-15-0
EINECS: 251-751-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR