D-tert-Leucin (H-D-Tle-OH)

Lieferant: Molekula

Synonyme: H-D-Tle-OH

45088529-250MG
MOLE45088529-250MGEA 0 EUR
MOLE45088529-250MG
D-tert-Leucin (H-D-Tle-OH)
D-tert-Leucin (H-D-Tle-OH)
Formel: C₆H₁₃NO₂
Molecular Weight: 131,2 g/mol
MDL: MFCD00004265
CAS-Nummer: 26782-71-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR