4-Methoxyphenylhydraziniumchlorid

Lieferant: Molekula

45290091-25G
MOLE45290091-25GEA 0 EUR
MOLE45290091-25G
4-Methoxyphenylhydraziniumchlorid
4-Methoxyphenylhydraziniumchlorid
Formel: C₇H₁₁ClN₂O
MDL: MFCD00012945
CAS-Nummer: 19501-58-7
EINECS: 243-115-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR