trans-4-Hydroxy-L(-)-prolin
Lieferant: Molekula
Synonyme:
L(-)-trans-4-Hydroxyprolin, (S)-(-)-trans-4-Hydroxyprolin, H-Hyp-OH, (2S)-(-)-trans-4-Hydroxypyrrolidin-2-carbonsäure, (S)-(-)-trans-4-Hydroxy-1,3,4,5-tetrahydro-2H-pyrrol- 2-carbonsäure, (S)-(-)-trans-4-Hydroxyazolidin-2-carbonsäure, (S)-(-)-trans-4-Hydroxypyrrolidin-2-carbonsäure
45298012-25G
MOLE45298012-25GEA
0
EUR
MOLE45298012-25G
trans-4-Hydroxy-L(-)-prolin
trans-4-Hydroxy-L(-)-prolin
Formel:
C₅H₉NO₃ Molecular Weight: 131.13 g/mol Siedepunkt: 296 Schmelzpunkt: 206 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064320 CAS-Nummer: 51-35-4 EINECS: 200-091-9 Merck Index: 13,04866 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...