(Benzotriazol-1-yloxy)tris(dimethylamino)phosphoniumhexafluorophosphat (BOP)
Lieferant: Molekula
Synonyme:
1H-Benzotriazol-1-yloxytris(dimethylamino)phosphonium hexafluorophosphate, ((1H-Benzo[d][1,2,3]triazol-1-yl)oxy)tris(dimethylamino)phosphonium hexafluorophosphate(V), BOP reagent, Benzotriazol-1-yloxytris(dimethylamino)-phosphonium hexafluorophosphate
45328655-25G
MOLE45328655-25GEA
117
EUR
MOLE45328655-25G
(Benzotriazol-1-yloxy)tris(dimethylamino)phosphoniumhexafluorophosphat (BOP)
(Benzotriazol-1-yloxy)tris(dimethylamino)phosphoniumhexafluorophosphat (BOP)
Formel:
C₁₂H₂₂F₆N₆OP₂ Molecular Weight: 442.28 g/mol |
MDL:
MFCD00011948 CAS-Nummer: 56602-33-6 EINECS: 260-279-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...