2,5-Dimercapto-1,3,4-thiadiazol
Lieferant: Molekula
Synonyme:
1,3,4-Thiadiazole-2,5-dithiol
45380441-100G
MOLE45380441-100GEA
0
EUR
MOLE45380441-100G
2,5-Dimercapto-1,3,4-thiadiazol
2,5-Dimercapto-1,3,4-thiadiazole
Formel:
C₂H₂N₂S₃ Molecular Weight: 150,25 g/mol |
MDL:
MFCD00003103 CAS-Nummer: 1072-71-5 EINECS: 214-014-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...