Tetraoctylammoniumbromid

Lieferant: Molekula

Synonyme: Tetra-n-octylammonium bromide

45634299-25G
MOLE45634299-25GEA 68.2 EUR
MOLE45634299-25G
Tetraoctylammoniumbromid
Tetraoctylammoniumbromid
Formel: C₃₂H₆₈BrN
Molecular Weight: 546,8 g/mol
Schmelzpunkt: ~ 96 °C
Flammpunkt: 100 °C
MDL: MFCD00011863
CAS-Nummer: 14866-33-2
EINECS: 238-936-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR