(2R)-2-Pyrrolidinylmethanol
Lieferant: Molekula
Synonyme:
D-Prolinol, (2R)-pyrrolidin-2-ylmethanol, D(-)-Prolinol
45777293-5G
MOLE45777293-5GEA
71.6
EUR
MOLE45777293-5G
(2R)-2-Pyrrolidinylmethanol
(R)-(-)-Prolinol
Formel:
C₅H₁₁NO Molecular Weight: 101,1 g/mol |
MDL:
MFCD00064321 CAS-Nummer: 68832-13-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...