2-Hydroxycarbazol

Lieferant: Molekula

Synonyme: 2-Carbazolol, Carbazol-2-ol

45893442-5G
MOLE45893442-5GEA 0 EUR
MOLE45893442-5G
2-Hydroxycarbazol
2-Hydroxycarbazole
Formel: C₁₂H₉NO
Molecular Weight: 183.21 g/mol
MDL: MFCD00004962
CAS-Nummer: 86-79-3
EINECS: 201-699-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR