N-Boc-glycin (Boc-Gly-OH)

Lieferant: Molekula

Synonyme: Boc-Gly-OH

46029276-25G
MOLE46029276-25GEA 116 EUR
MOLE46029276-25G
N-Boc-glycin (Boc-Gly-OH)
N-Boc-glycin (Boc-Gly-OH)
Formel: C₇H₁₃NO₄
Molecular Weight: 175,2 g/mol
MDL: MFCD00002690
CAS-Nummer: 4530-20-5
EINECS: 224-864-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR